Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

ProKlenz® TWO High Performance Acid Detergent

Supplier: Steris
ProKlenz®
142508 142201WR 142205WR 142501 142505 1425D3
77531-600CS 327.07 USD InStock
77531-600 11005-681 76191-347 CA11005-678 11005-679 CA11005-680
ProKlenz® TWO High Performance Acid Detergent
Cleaning Agents Detergents and Surface Cleansers
For routine cleaning, derouging and passivation.

  • Productivity - Effective for routine removal of inorganic soils and rouge as well as for passivation in clean-in-place (CIP), clean-out-of-place (COP), and manual applications
  • Easy to validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Cost savings - Reduces water consumption and cleaning time due to superior rinsing and low foaming characteristics
  • Globally availability - phosphate-free, and manufactured with biodegradable ingredients to support harmonization of cleaning processes
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility

ProKlenz® TWO Acid detergent is a proprietary blend of organic acids, an advanced surfactant system and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming product removes a wide range of process residues, inorganic soils and rouge, and is ideal for use in CIP, COP, manual and passivation applications.

ProKlenz® TWO High performance acid detergent is a specially formulated phosphate-free for routine cleaning, derouging and passivation in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective validatable detergent.

CAS:: 77-92-9; 144-62-7
UN 3265
Hazard class: 8
Packing group: III

Caution: Danger, GHS05.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty