Would you like to log in again?
Stainless Steel Calibration Weights, ASTM, Class 1, Ohaus®
Supplier: Ohaus
80780025 80780029 80780021 80780022 80780028 80780023 80780026 80780020 80780024 80780027
11384-120EA 783.74 USD InStock
11384-120 11384-130 11384-122 11384-132 11384-124 11384-134 11384-126 11384-136 11384-128 11384-138
Stainless Steel Calibration Weights, ASTM, Class 1, Ohaus®
Weights Calibration Weights
Manufactured to international standards, individual calibration weights are available for a variety of precision applications
Calibrated to ASTM Class 1. Extra-fine accuracy for calibrating high precision analytical balances with a readability as low as 0.1 to 0.01mg.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty