Would you like to log in again?
Individual Precision Analytical Weights, Class 2, Troemner
Supplier: Troemner
80858259 80858288 80858287 80858301 80858368 7014-2T 7014-2 80858404 80858328 80858443 80858226 80858183 80858228 80858638 7016-2 80858701 80858611 80858613 80858406 80858637 80858181 7016-2W 7012-2T 80858227 7016-2T 80858198 80858666 80858442 80858534 80858331 80858506 80858213 80858403 80858272 80858504 7013-2T 80858303 80858371 80858663 80858481 7014-2W 80858256 80858483 80858258 80858197 80858273 80858636 80858480 80858574 80858199 80858124 80858572 80858369 80858302 80858532 80858530 80858703 80858241 80858212 7010-2W 80858329 80858242 7010-2T 80858664 80858705 80858286 80858445 80858274 80858505 7013-2W 80858610 80858182 80858240 7012-2W 80858214 7010-2 80858569
12666-066EA 77.78 USD InStock
12666-066 12666-188 12666-100 12666-068 12666-102 12666-104 14216-786 12666-106 14216-788 12666-180 12666-060 12666-182 12666-062 12666-184 12666-140 12666-064 12666-186 12666-154 14216-772 12666-110 12666-078 12666-156 14216-774 12666-112 12666-114 12666-158 14216-776 12666-116 12666-190 12666-070 12666-072 12666-150 12666-074 12666-152 12666-076 12666-108 12666-088 14216-760 12666-122 12666-166 14216-762 12666-168 12666-124 12666-126 14216-764 14216-800 12666-128 12666-080 12666-082 12666-160 12666-084 12666-162 12666-086 12666-164 12666-120 12666-118 12666-176 14216-750 12666-132 12666-056 14216-752 12666-134 12666-178 14216-796 12666-058 12666-136 14216-798 12666-138 12666-170 12666-050 12666-172 12666-052 12666-174 12666-130 12666-054 12666-090 14216-748
Individual Precision Analytical Weights, Class 2, Troemner
Weights Calibration Weights
Highly polished kilogram and gram weights made of Troemner Alloy 8, which is a specially developed austenitic stainless steel that possesses closely controlled density, extremely low magnetic permeability, good stability, and resistance to corrosion and handling damage
Fractional weights 500 mg to 5 mg are made of stainless steel (30 mg weights are aluminum); fractional weights 3 mg and below are made of aluminum.
This exclusive material has a density of 8.0 at 25°C (controlled to ±0.1%), is essentially nonmagnetic in the annealed condition, and will not transform to magnetic under any combination of temperature and cold work down to –195.5°C (–320°F).
Certifications: Where applicable, weights are supplied with a NIST/NVLAP Weight Calibration Certificate or a Traceable Certificate.
Order Now
ORDER
SERVICES
Services: VWR Catalyst™ Services provides convenient and fast re-calibration services within 6 business days for your weights. Please contact us at 1.888.793.2300 or VWRCATALYST@vwr.com for a quote or more information. If your weights are found to be out of tolerance, we will make the proper adjustments to bring them back into tolerance. To prevent loss and ensure the timely processing and integrity of your precision weights and/or calibration devices, please do not pack in foam peanuts. Please avoid packing your items with foam, packing peanuts or other material that cannot be recycled.
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty