Precision-crafted of stainless steel, with a highly polished finish.
Weights 30kg to 5kg are flat on top with a recessed grip handle for easy handling and stacking
Weights 5kg and below supplied in a rugged, precision-fit, high quality polycarbonate case with handling glove.
Weights 5kg to 1kg are cylindrical in shape with a groove near the top to allow for easy handling and gripping. Weights below 1kg are straight cylinders. All are manufactured with adjusting cavities.
Certifications: Where applicable, weights are supplied with a NIST/NVLAP Weight Calibration Certificate or a Traceable Certificate.
Ordering information: To order cases for weights above 5kg, and to order ANSI/ASTM Classes 2 and 3 weights, contact your Avantor sales representative.