Would you like to log in again?
Nalgene® Griffin Low Form Graduated Beakers, Polypropylene, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
1201-1000 1201-0400 1201-0050 1201-0600 1201-4000 1201-2000 1201-0150 1201-0030 1201-0100 1201-0250 1201-0400
13915-090CS 326.58 USD InStock
13915-090 13915-180 CATX13915-114 13915-147 13915-114 13915-125 13915-169 13915-103 13915-078 13915-067 13915-089
Nalgene® Griffin Low Form Graduated Beakers, Polypropylene, Thermo Scientific
Beakers
Heavy-duty, translucent polypropylene beakers with ribs for stacking.
  • Easy-to-read graduations
  • Autoclavable
  • Chemically resistant
Beakers feature easy-to-read graduations with size and resin codes. Maximum use temperature and no-flame symbol printed in black. Autoclavable and chemically resistant. Not for use on a hot plate.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty