Would you like to log in again?
STEEL-BRIGHT, Veltek Associates
Supplier: Veltek
STEEL-BRIGHT®
SB-02
14003-342CS 820.21 USD InStock
14003-342
STEEL-BRIGHT, Veltek Associates
Cleaning Agents Detergents and Surface Cleansers
Designed to remove chemical residues, spotting, and staining on stainless steel, chrome, brass, aluminum, and copper.
This sterile cleaning agent will not rainbow, accumulate a heavy build-up, or leave a powdery residue. Filtered at 0.2µm. Gamma-irradiated, double-bagged, and delivered with lot-specific sterile documentation.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty