Would you like to log in again?
DA-7645® Process and Research Cleaner, Steris
Supplier: Steris
DA-7645®
116501 1165D3 116505
77531-102EA 14668.5 USD InStock
77531-102 14004-166 14004-168
DA-7645® Process and Research Cleaner, Steris
Cleaning Agents Detergents and Surface Cleansers
DA-7645® is a general-purpose cleaner, detergent is a proprietary blend of surfactants, solvents and other performance-enhancing ingredients that provide multiple cleaning mechanisms in manual cleaning applications. It can also be used as a detergent additive to boost the cleaning performance of alkaline detergent systems.
  • Efficacy - Mild pH formulation delivers excellent cleaning characteristics and substrate compatibility with enhanced operator safety
  • Easy to Validate - Facilitates cleaning validation with a comprehensive support documentation package including specific and non-specific analytical methods, toxicity, and substrate compatibility reports
  • Productivity - Delivers efficiency to manual cleaning operations using a blend of surfactants, solvents, and other performance-enhancing ingredients
  • Reliability - Manufactured for the pharmaceutical industry in a cGMP compliant, FDA registered and ISO 9001 certified facility
Provides manual cleaning with enhanced substrate compatibility in the pharmaceutical, biotechnology, cosmetic, dietary supplement, and other industries requiring an effective detergent.
A broad variety of tough-to-clean soil types, including ointments, creams, waxes and petrolatum-based residues, are readily removed with DA-7645®
Ordering information: .
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty