Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

CIP 150® Alkaline Process Cleaning Detergent

Supplier: Steris
CIP 150®
1D15D3 1D1505 1D1502 1D1505 1D1501
77531-106EA 1819.12 USD InStock
77531-106 77531-098 14004-232 CA14004-232 14004-230
CIP 150® Alkaline Process Cleaning Detergent
Cleaning Agents Detergents and Surface Cleansers
Process cleaning detergent For CIP, COP and manual applications.

CIP 150® Alkaline detergent is a proprietary blend of potassium hydroxide, sodium hypochlorite (oxidizing agent), surfactants and other performance-enhancing ingredients that provide multiple cleaning mechanisms. This low foaming product removes a wide range of process residues, from fermentation by-products to silicone-based emulsions and lubricants, and is ideal for use in CIP, COP and manual applications.

CIP 150® Alkaline process and research cleaner is specially formulated to meet the unique cleaning demands found in biopharmaceutical manufacturing. The blend of alkalinity, surfactants, and oxidizing agents were developed for the most challenging protein-based process residues.

CAS: 1310-58-3; 7681-52-9
UN 3266
Hazard class: 8
Packing group: III

Caution: Danger, GHS05; GHS07.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty