Would you like to log in again?
SP Wilmad-LabGlass 4-Neck Molded Reaction Vessel Covers, SP Industries
Supplier: SP Industries
LG-8081-100 LG-8083-100
14205-508EA 1149.64 USD InStock
14205-508 14204-494
SP Wilmad-LabGlass 4-Neck Molded Reaction Vessel Covers, SP Industries
Lids Reaction Vessel Lids
These borosilicate glass 4-neck reaction vessel lids/covers have a contact surface that is finely ground to provide a tight seal.
  • LG-8081 is designed for use with LG-8080 cylindrical reaction vessels
  • LG-8083 is designed for use with LG-8082 vessels
Featuring a flat flange, these pressed lids have a center neck and three side necks with outer standard taper joints. Use LG-7316 clamps.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty