Would you like to log in again?
Due to maintenance activity, Global web site will not be available from 6AM till 2PM ET on 20th April 2024.
SMITH & WESSON® MAGNUM 3G™ Safety Glasses KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
Smith & Wesson®
19940 19829 20337 19830 19823 19793 19792 19794 19961 19955 19824 19799 19822
14219-778EA 23.94 USD InStock
14219-778 14219-788 14219-776 67200-202 67200-208 14219-782 14219-760 14219-770 14219-780 14219-764 14219-786 14219-762 14219-772
SMITH & WESSON® MAGNUM 3G™ Safety Glasses KIMBERLY-CLARK PROFESSIONAL®
Safety Glasses
Safety glasses are ergonomically designed to provide optimal comfort and protection.
  • All lenses provide 99.9% UV protection
  • #3 filter lens coating is ideal for welding sites and for light brazing or cutting
  • #5 filter lens is also ideal for welding sites and for heavy brazing or cutting
All lenses provide 99.9% UV protection. The #3 filter lens coating is ideal for welding sites and for light brazing or cutting. The #5 filter lens is also ideal for welding sites and for heavy brazing or cutting.
Glasses feature a 7-base, cut-to-shape spherical UV blocker lens. Nose pad posts are molded directly onto the lens piece, and retention screws have been replaced with a mechanical snap fit of lens to frame. A built-in hinge eliminates scratching of lens interior. All features aid in ensuring lightweight, secure eye protection. The Mini-Magnum models are equipped with all of the same features on smaller frames for younger wearers.
Certifications: Glasses exceed ANSI Z87.1 standards.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty