Would you like to log in again?
Nalgene® Boston Round Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2002-0008 2002-0001 2002-9025 2004-9050 2002-9050 2002-9016 2002-0006 2002-0002 2002-0004 2004-0002 2004-0001 2002-0032 2004-0008 2004-9125 2004-0016 2004-0004 2004-0032 2002-9125 2004-9025 2002-0016
16057-110CS 353.28 USD InStock
16057-110 16061-966 16061-988 16057-029 16057-007 16057-109 16062-109 16062-007 16062-029 16062-120 16062-040 16062-084 16062-062 16056-988 16056-966 16057-084 16057-040 16057-062 16057-142 16057-120
Nalgene® Boston Round Bottles, High-Density Polyethylene, Narrow Mouth, Thermo Scientific
Bottles Plastic Bottles
Nalgene® Lab Quality Bottles are specifically designed for demanding lab use.
  • More rigid than LDPE
  • Excellent chemical resistance, even with most corrosives
  • Suitable for freezer use to –100 °C (–148 °F)
These leakproof, translucent, all-purpose Boston Round bottles are highly reliable and durable for long-term use
Manufactured from validated and controlled resins meeting a variety of food and pharmaceutical regulatory requirements.
They are suitable for everything from general lab use to storing critical, high value materials. The narrow mouth is ideal for storing, shipping, and packaging liquids.
Ordering information: For replacement closures for 16057-110 and 16057-142, see 16219-122 series.
Delivery information: Bottles are supplied with polypropylene screw closures.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty