Would you like to log in again?
Nalgene® Drop-Dispenser Bottles, Low-Density Polyethylene, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
2411-0125 2411-0030 2411-0250 2411-0060 2411-0015
16354-465CS 403.2 USD InStock
16354-465 16354-421 16354-487 16354-443 16354-400
Nalgene® Drop-Dispenser Bottles, Low-Density Polyethylene, Thermo Scientific
Bottles Dropping and Dispensing Bottles
Bottles dispense one drop at a time.
Captive spout cap won't release or leak when bottle is squeezed.
Delivery information: Supplied with polypropylene dropping closure and spout plug.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty