Would you like to log in again?
VWR® Adjustable Spray Wash Bottle
Supplier: VWR
VWR®
90027-53
16650-049EA 89.03 USD InStock
16650-049
VWR® Adjustable Spray Wash Bottle
Bottles Wash Bottles
This double-headed wash bottle has an adjustable-spray stream and will keep contents completely free of contaminants if the sprayer and side-fill opening are in the closed position.
  • Convenient double-headed design and side-fill opening
  • Instant-off release eliminates drips
  • High-density polyethylene construction
The convenient side-fill opening helps to maintain the purity of solutions. Large capacity minimizes refilling. Adjustable spray pattern is controlled by a contoured spray trigger with 'instant off.'Virtually closed system eliminates drips and handles hot, cold, aqueous, and solvent-based solutions. Engraved markings show liquid amount at a glance. High-density polyethylene bottle with polypropylene sprayer.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty