Would you like to log in again?
PRISM® 401™ Instant Adhesive, Surface Insensitive, Loctite®, Henkel
Supplier: Henkel
PRISM® 401™
233641 135429 135429
19807-532CS 6302.09 USD InStock
19807-532 19807-530 37001-742
PRISM® 401™ Instant Adhesive, Surface Insensitive, Loctite®, Henkel
Adhesives
This surface-insensitive adhesive is ideal for difficult-to-bond substrates.
  • Ideal for close fitting applications
  • 3,200 psi tensile strength
  • Fills gaps up to 0.012 cm (0.005")
  • Agriculture Canada approved
Adhesive is ideal for use with rough, porous, and acidic surfaces, such as wood, paper, leather, or fabric.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty