Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MEK1 Rabbit Polyclonal Antibody

Supplier: Enzo Life Sciences
KAP-MA010E KAP-MA010-E
200062-046EA 406 USD InStock
200062-046 CA200062-046
Anti-MEK1 Rabbit Polyclonal Antibody
Antibodies
MEK is a dual specificity kinase capable of phosphorylating both tyrosine and threonine residues. The MEK family, which is also known as MAP kinase kinase, phosphorylates MAP kinases on the conserved T-X-Y motif; phosphorylation of MAPK by MEK results in an increase in MAPK activity. MEK is involved in a diverse array of cellular processes such as stress-activated response, apoptosis, cytokine-induced cell proliferation, and DNA recombination during meiosis.

  • For Immunofluorescence, Immunohistochemistry, Western Blot

Recognizes human, mouse, rat and Xenopus MEK1. Detect a band of ~48kDa by Western blot.

Type: Primary
Antigen: MEK1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty