Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Safe-Lock Tubes, 0.5 ml - Microtubes

Supplier: Eppendorf
Eppendorf®
022363662 022600001 022363662 022363638 022363646 022363638 022600001 0030123603 022363611 022363719 022363654 022363697 022363611 022363671 022363697 022363654 022363719 022363671 022363646
20901-505CS 95.11 USD InStock
20901-505 CA20901-548 CA20901-515 20901-515 CA20901-505 20901-548 CA20901-535 CA20901-525 76019-012 20901-510 20901-520 20901-525 CA20901-520 CA20901-510 20901-535 CA53511-996 53511-996 62111-750 CA62111-750
Safe-Lock Tubes, 0.5 ml - Microtubes
Tubes Microcentrifuge Tubes
Eppendorf® Safe-Lock tubes 0.5 ml - Microtube ensures secure sample storage. With a unique locking mechanism, these tubes prevent accidental opening, safeguarding precious samples. Ideal for PCR, these tubes offer reliability and ease in a compact design.

  • Hinged Safe-Lock lid prevents unintended opening during incubation and storage
  • g-Safe® technology, dependent on the Safe-Lock tube variant (refer to the manual), ensures exceptional centrifugation stability up to 30000×g
  • g-Safe® technology prevents sample loss in the event of tube breakage, and provides enhanced safety when handling hazardous samples
  • Polypropylene construction offers high resistance to chemicals, mechanical stress, and extreme temperatures
  • Precise lid sealing minimizes evaporation rates during long-term storage
  • Large frosted lid and side surface facilitate easy labeling
  • Ensured functionality in a temperature range from −86 to +100 °C
  • Autoclavable when open (121 °C, 20 min.) for effective sterilization
  • Available in lot-certified PCR Clean, Eppendorf Forensic DNA Grade, and Biopur® quality, ensuring tested purity

Eppendorf® Safe-Lock 0.5 ml microtubes epitomize advanced sample storage, featuring a patented Safe-Lock design for unparalleled security. Tailored for PCR applications, these microtubes provide a space-efficient 0.5 ml capacity, ideal for labs with limited storage. Their compatibility with various centrifuge models ensures seamless integration into existing workflows. Robust construction and user-friendly attributes, such as clear graduations, further enhance reliability and ease of handling. Elevate laboratory practices with Eppendorf® Safe-Lock tubes, which set a new standard for sample integrity and operational efficiency.

Manufactured without the use of slip agents, plasticizers, and biocides – substances that have been shown to leach from plastic consumables into the sample and negatively affect bio-assay results.

Certifications: Certificate of Quality/Conformity, Lot-specific certficates for the purity grades PCR clean, Biopur®, Forensic DNA grade can be downloaded from the Eppendorf website.

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty