Would you like to log in again?
High-Strength Round-Bottom Centrifuge Tubes, Glass, Kimble Chase
Supplier: DWK Life Sciences
45600-30 45600-15 45500-15 45500-30
46100-832CS 364.4 USD InStock
46100-832 46100-824 21021-424 21021-422
High-Strength Round-Bottom Centrifuge Tubes, Glass, Kimble Chase
Tubes Centrifuge Tubes
High strength 15 and 30 ml reusable centrifuge tubes are manufactured from Type 1, Class B, chemically strengthened borosilicate glass.
  • Withstand temperatures upto 300 °C (572 °F)
  • Screw-thread tubes are supplied with PTFE-faced rubber liner caps
These tubes can be centrifuged upto 13100 ×g when used with the accessory rubber adapter sleeve in a 50 ml rotor cavity.
Ordering information: Accessory sleeves and replacement caps sold separately.
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty