Would you like to log in again?
OPTI-SOLV® Biocompatible Solvent Reservoir Filter
Supplier: Optimize Technologies
OPTI-SOLV®
10-04-00053 10-04-00081 10-04-00080 10-04-00079 10-04-00047 10-04-00082 10-04-00051 10-04-00050
77100-048EA 210.63 USD InStock
77100-048 21510-496 21510-498 77100-050 77100-044 21510-492 77100-046 21510-494
OPTI-SOLV® Biocompatible Solvent Reservoir Filter
Filters Liquid Process Filters
A solid PTFE body keeps OPTI-SOLV® filter at the base of the bottle where a bottom-mounted titanium frit provides unrestricted solvent access.
  • Solid PTFE body keeps OPTI-SOLV filter at the base of the bottle
  • Bottom-mounted titanium frit provides unrestricted solvent access
  • Smooth flow path and conical design does not trap air
  • No tools or fittings required
As always, optimize makes installation as simple as possible - no tools or fittings required. OPTI-SOLV reservoir filters employ a unique conical design inside the filter housing to prevent air bubbles from getting trapped and disturbing your analysis. While the availability of HPLC-grade solvents diminishes the need for pre-filtering, it is still imperative that a filtration device be installed in the reservoir. Placing a filter in the reservoir is an economical way to filter particles that may result from buffer salt precipitation, airborne dust, improperly cleaned glassware, or microbial contamination; none of which are remedied by the use of HPLC-grade solvents.
Delivery information: Includes 5 feet of pre-installed PTFE tubing.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty