Would you like to log in again?
Coverage Plus NPD® One-Step Cleaner Disinfectants
Supplier: Steris
Coverage Plus NPD®
638701 638708
21899-844CS 559.2 USD InStock
21899-844 89126-752
Coverage Plus NPD® One-Step Cleaner Disinfectants
Cleaning Agents Disinfectants
STERIS' Coverage Plus NPD® One-Step Cleaner disinfectant is a liquid concentrate specifically formulated for the disinfection of hard surfaces. The use-dilution can be applied manually or sprayed.
  • Efficacy - Quaternary ammonium active ingredients provide broad spectrum antimicrobial efficacy
  • Productivity - Formulation includes surfactants for surface cleaning while disinfecting
  • Ease of Use - Ideal for use in a contamination control rotation
  • Reliability - Complete lot traceability
  • Assurance - Each lot documented by Certificate of Analysis
STERIS' Coverage Plus NPD® One-Step Cleaner disinfectant a phosphate-free, one-step cleaner/disinfectant formulated for use on hard non-porous surfaces. This product consists of four quaternary ammonium compounds formulated with a compatible detergent system. It is proven effective in the presence of heavy soil (5% bovine serum) and hard water. Coverage Plus NPD® disinfectant contains no perfumes or dyes that could interfere with critical research studies or manufacturing processes. Use-dilution is 1:256.
Caution: Danger, GHS02; GHS05; GHS07.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty