Would you like to log in again?
Super Polx® 1500 Wipers, Berkshire
Supplier: Berkshire
Super Polx®
S15000909.8 S1500.0909B6
21914-470CS 1058.98 USD InStock
21914-470 21914-472
Super Polx® 1500 Wipers, Berkshire
Wipes Cleanroom Wipes Dry Cleanroom Wipes
For use on soft or rough surfaces where particles and ion content are a concern.
  • Low particles, fibers, ions, and extractables
  • High abrasion resistance
  • Chemically compatible with IPA, acetone, and other solvents
  • Lightweight, high sorbency design
  • Low endotoxin levels
  • Soft hand and edge for sensitive surfaces
These all-purpose wipers are made from a continuous-filament, non-run 100% polyester knit that is abrasion- and chemical-resistant. Knitted construction promotes sorbency - wipers will absorb twice their weight.
Certifications: Laundered and packaged in an ISO Class 4 (FED-STD-209E Class 10/M2.5) cleanroom.
Packaging: Double-bagged to ensure cleanliness.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty