Would you like to log in again?
Syringe filters, Acrodisc®, Supor® membrane, Cytiva (Formerly Pall Lab)
Supplier: Cytiva
Acrodisc®
4661 4659 4668 4506 AP-4426 4662 4651 4650 4508 4661 4656 4655 4509 4602 4509 4504 4692 4653 4692 AP-4798 4506 AP-4799 4668 4658 4614 4508 4652 4612 4612 AP-4425 4656 4651 4608 4611 4611 4653 4614 4504 4654 4618 4660 4662 4655 4509 4650 AP-4425 4602 4604 4525 4660 AP-4426 4608 4187 4187 4652 4604 4659 4658 AP-4799 4525 4618 4654
28150-956PK 177.79 USD InStock
28150-956 28144-009 34181-088 34181-086 CA28139-710 28139-702 28150-958 28150-957 CA28143-372 CA28143-295 28143-044 CA28143-374 28143-046 CA28143-301 CA28143-300 CA28143-303 CA28143-302 CA28143-348 CA28143-304 CA28143-309 28143-372 28143-295 28143-374 CA28143-310 28143-300 28143-301 CA28143-312 28143-302 28143-303 28143-304 CA28143-314 28143-305 CA34181-078 CA97035-174 CA97035-176 28143-309 97027-954 97027-952 CA28143-350 CA28143-352 CA28143-046 28143-312 CA28143-048 CA28143-400 28143-314 CA34181-086 CA34181-088 CA28144-009 28139-710 34181-078 CA28150-958 CA28150-956 CA97027-954 97035-178 97035-176 97035-174 28143-350 28143-352 CA28143-044 28143-310
Syringe filters, Acrodisc®, Supor® membrane, Cytiva (Formerly Pall Lab)
Filters Syringe Filters
Acrodisc™ syringe filters with Supor™ membrane deliver high flow rates and low protein binding.
  • Superior flow rates and higher throughputs than competitive devices
  • Low protein binding to minimize sample loss
  • Pre-sterilized products are sterilized by gamma irradiation to eliminate potential contamination by EtO residuals
  • A range of sizes (13 to 37 mm) accommodates sample volumes from < 10 to 150 ml
  • Acrodisc PF and Serum Acrodisc syringe filters feature built-in prefilter for increased throughput of difficult-to-filter liquids
  • Reduces mycoplasma with the use of 0,1 μm pore size
Supor™ hydrophilic polyethersulfone (PES) membrane: PP housing (except 25 and 32 mm: Acrylic and Acrodisc Serum: ABS)
Applications:
0.1 and 0.2 μm pore sizes provide sterilization of small volumes of buffers, culture media, and additives.
Acrodisc PF and Serum Acrodisc syringe filters are ideal for clarification/sterilization of viscous or particulate-laden solutions.
Use larger pore size filters for prefiltration and particulate removal.
Certifications: Passes United States Pharmacopeia (USP) Biological Reactivity Test, In Vivo <88>.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty