Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Acrodisc® Syringe Filters, 25 mm, Cytiva (Formerly Pall Lab)

Supplier: Cytiva
Acrodisc®
4503 4914 4438 4522 4219-N 4438TC 4521 4502 4521 4225 4408TC 4226 4520 4927 4927 4436 4501 4408 4929 4528 4219-N 4520 4521 4528 4436 4914 4501 4225 4436 4438 4438TC 4219-N 4408 4438 4406TC 4226 4406TC 4406 4406 4225 4502 4503 4225 4500 4521 4500 4406 4408 4436 4219-N 4549 4408 4932 4932 4408TC 4549 4406 4522 4438
76308-640PK 1867.7 USD InStock
76308-640 28143-924 28143-926 CA28143-936 28139-304 77603-460 28143-928 CA77603-474 77603-462 CA77603-472 77603-456 CA76894-086 28143-280 28143-286 CA76894-088 76308-634 CA77007-786 CA28143-940 CA76308-640 28143-936 CA28143-946 CA77603-462 76308-636 CA28139-304 CA28143-948 76308-638 77603-454 CA28144-584 CA77603-460 CA28144-582 CA77603-456 76894-088 CA28144-580 CA77603-454 76894-086 77007-786 CA28100-062 28143-940 CA76308-638 28143-946 28143-948 77603-482 CA76308-634 28144-588 28144-583 28144-584 CA28144-592 CA28144-591 CA28144-590 28100-062 CA28143-249 77603-472 CA28143-924 77603-474 CA28143-926 CA77603-482 CA28143-928 28144-592 28144-593
Acrodisc® Syringe Filters, 25 mm, Cytiva (Formerly Pall Lab)
Filters Syringe Filters
These 25 mm syringe filters offer an available prefiltration layer and wide choice of filter media.

  • Wide range of filter media available
  • Available in 0.2 µm for UHPLC applications
  • Available with pre-filter for difficult to filter samples
  • Color coded for easy identification

Polypropylene housing with female Luer inlet and male slip outlet connections. Some models feature a GxF borosilicate glass pre-filter, which provides two to four times the throughput of standard glass fiber prefilter devices, allowing for quick and easy filtration of difficult-to-filter samples.

Available media options:
1. NEW UNIVERSAL wwPTFE (water wettable) membrane has wide chemical compatibility, fast flow rates, low protein binding, and low levels of UV-absorbing extractables.
2. PVDF membrane has excellent compatibility with aggressive acids and alcohols.
3. PES (hydrophilic polyethersulfone) membrane is available in a special ion Chromatography design or with a GxF prefilter.
4. Hydrophilic Nylon membrane has excellent compatibility with esters, bases, and alcohols, and requires no prewetting.
5. Hydrophobic PTFE membrane with polypropylene support offers exceptional chemical and temperature compatibility and is ideal for gas, organic solutions, air filtration, or venting.

For ion chromatography specific syringe filters, look for the Acrodisc® Ion Chromatography Syringe Filters page.

Certifications: All membranes are HPLC certified for low levels of UV-absorbing extractables.

Order Now
 

ORDER

SPECIFICATIONS

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty