Would you like to log in again?
Whatman™ Autovial Syringeless Filters
Supplier: Whatman products (Cytiva)
Whatman™ Autovial™ Autovial™
AV125ENAO AV125EAQU AV115NPUORG AV115NPUNYL AV125UCA AV525UAQU AV125UNAO AV125UGMF AV125EORG AV115UGMF AV525UORG AV125UNAO AV125UAQU AV115NPUORG AV125EAQU AV115NPUAQU AV125NPUPSU AV125UORG AV115NPEORG AV125EPP AV125NPUAQU AV525UNAO AV125UAQU AV125UORG AV115UGMF AV125UPP AV125EORG AV115NPUNYL AV125EPP AV125NPUAQU AV125ENAO AV525UORG AV125NPUPSU AV115NPUAQU AV525UNAO AV125UPP AV125UCA AV115NPUAQU AV115NPEORG AV125UGMF
28297-108PK 448.96 USD InStock
28297-108 CA28296-030 80089-522 28297-106 28297-104 CA28296-012 28297-004 28296-024 28297-124 CA28297-118 CA28297-030 CA28297-032 28297-122 28297-022 21426-500 28296-028 CA11014-020 28297-120 21426-503 10035-852 CA28297-108 28296-030 28297-118 CA10147-472 28296-012 CA80089-522 CA28297-122 CA28297-124 CA28297-004 CA28297-104 CA28297-106 AP400001-986 CA28297-120 CA28297-022 CA28296-024 CA21426-503 10192-070 28297-032 CA28296-028 28297-030
Whatman™ Autovial Syringeless Filters
Filters Syringeless Filters
The Whatman Autovial Syringeless Filter replaces multiple syringe-coupled sample filtration devices with a single, disposable unit.
  • Available in 5 or 12 ml capacity
  • Single unit convenience – pre-assembled and easy to load
  • Self-contained, preventing filter pop-off with hazardous samples
  • Range of sterile or nonsterile membrane media available
  • Built-in air purge maximizes sample recovery
  • Glass fiber or polypropylene prefilter in selected 12 ml models – for difficult-to-filter samples
  • Choice of glass fiber or polypropylene prefilters
Autovial from Cytiva's business combines Whatman quality membranes and devices with an all-in-one syringeless filter design.
Filtration involving multiple consumables like syringe filters, vials, and septa increases storage space required for these components and time required to perform sample filtration. Cytiva's Autovial combines all of these components into a single consumable.
Autovial devices are comprised of two parts: a graduated filter barrel and a plunger. Sample is poured into the filter barrel, and the plunger is inserted until the bottom is securely in place. The tip of the device is placed into the mouth of an autosampler vial and the plunger compressed.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty