These borosilicate glass filters are ideal for measuring landfill leachate composition with a TCLP test. They can also be used for DNA binding and purification, filtering finely precipitated proteins and filtering biochemical solutions, fluids, and nucleic acids.
- Very fine particle retention
- Good loading capacity and flow rate
- Binder free