Would you like to log in again?
Weiler® Crimped Wire Solid End Brushes, ORS Nasco
Supplier: Weiler
804-10014 804-10006 804-10004 804-10024 804-10012 804-10007 804-10018 804-10015 804-10158 804-10008 804-10128 804-10013 804-10010 804-10020 804-10003 804-10023 804-10001 804-10009 804-10002 804-10016 804-10011 804-10017 804-10022 804-10005 804-10019 804-10021
300048-440EA 18.97 USD InStock
300048-440 300048-442 300087-840 300010-150 300010-151 300010-152 300046-678 300010-153 300010-154 300010-144 300010-145 300010-146 300010-147 300010-148 300010-149 300046-680 300048-434 300048-436 300046-688 300048-438 300010-143 300010-155 300047-700 300047-704 300047-702 300091-882
Weiler® Crimped Wire Solid End Brushes, ORS Nasco
Brushes
Crimped brushes are ideal for general-purpose and light duty applications to remove rust, paint, oxides, and scale.
  • Light-duty cleaning in corners and hard to reach areas
  • Solid end construction
  • Very flexible brushing action
Use with power drills, drill presses, die grinders, and right-angle air die grinders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty