Would you like to log in again?
Weiler® Heavy-Duty Knot Wire Cup Brush, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-12556 804-12766 804-12926 804-12916 804-12756
300010-166EA 83.48 USD InStock
300010-166 300042-084 300042-158 300042-156 300010-165
Weiler® Heavy-Duty Knot Wire Cup Brush, ORS Nasco
Brushes
Knot wire cup brushes provide very aggressive brushing action while still maintaining some flexibility.
  • Best suited for demanding cleaning applications on large surface areas
  • Ideal tool for severe cleaning applications
These knot brushes are designed for use with right-angle grinders to remove rust, paint, oxides, and scale.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty