Would you like to log in again?
Proto® TorquePlus™ Sockets, ORS Nasco
Supplier: ORS Nasco
577-5566 577-5538 577-5432H 577-5530 577-5552 577-5442 577-5776 577-5424H 577-5796 577-5426 577-5414H 577-5540 577-5546 577-5544 577-5764 577-5558 577-5428H 577-5758 577-5424 577-5430H 577-5434 577-5738 577-5572 577-5788 577-5556 577-5570 577-5428 577-5754 577-5766 577-5782 577-5550 577-5748 577-5440 577-5412H 577-5438H 577-5524 577-5768 577-5800 577-5568 577-5446H 577-5536 577-5410 577-5554 577-5416 577-5770 577-5422 577-5542 577-5420 577-5418 577-5412 577-5564 577-5756 577-5420H 577-5434H 577-5444 577-5416H 577-5438 577-5560 577-5740 577-5431 577-5746 577-5444H 577-5446 577-5436H 577-5760 577-5532 577-5430 577-5436 577-5794 577-5548 577-5534 577-5448H 577-5734 577-5432 577-5780 577-5528 577-5736 577-5772 577-5440H 577-5526 577-5744 577-5426H 577-5576 577-5422H 577-5442H 577-5784 577-5812 577-5752 577-5418H 577-5414 577-5448 577-5742
300027-146EA 170.93 USD InStock
300027-146 300027-026 300027-148 300026-818 300027-140 300027-064 300027-142 300027-022 300027-066 300027-144 300026-896 300026-610 300026-976 300027-070 300026-856 300026-772 300026-850 300026-696 300027-036 300027-158 300027-114 300027-116 300027-118 300026-806 300027-150 300027-074 300026-728 300027-152 300027-076 300027-154 300027-078 300027-112 300027-156 300026-840 300026-802 300026-648 300026-846 300027-080 300026-844 300026-768 300027-082 300026-680 300026-640 300026-684 300027-124 300027-004 300027-126 300027-160 300027-040 300027-084 300027-162 300026-914 300027-164 300003-018 300027-120 300026-918 300027-122 300027-166 300027-046 300026-996 300026-832 300026-752 300002-966 300026-912 300026-836 300002-968 300026-712 300026-756 300026-790 300027-058 300027-136 300027-016 300027-138 300027-050 300026-828 300026-906 300026-826 300027-130 300027-054 300027-010 300027-132 300027-134 300026-908 300026-700 300026-664 300026-786 300026-824 300026-902 300026-668 300026-988 300026-900 300026-982
Proto® TorquePlus™ Sockets, ORS Nasco
Tool Sets
The TorquePlus™ internal design minimizes fastener rounding, helps to extend fastener life, and allows the user to more easily turn worn, rounded, or rusted fasteners.
Proto® Sockets are ideal for use on stubborn or damaged fasteners, and also on fasteners made from soft materials. The TorquePlus™ design is a high performance drive configuration which allows for quick and easy socket engagement to the fastener flats, not the corners. Measurements are provided in inches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty