Would you like to log in again?
Portable Roll Groovers, Ridgid®, ORS Nasco
Supplier: ORS Nasco
632-88232
300034-240EA 1659.39 USD InStock
300034-240
Portable Roll Groovers, Ridgid®, ORS Nasco
Tool Sets
Optional Roll Sets for: Copper 2 to 6" (Types K, L, M, DWV) 1". 10, 40 steel pipe. Optional ¼ to 1½" and 2 to 6" AWWA Roll Sets are also available.
  • Quick and easy mount to RIDGID Nos. 300 Power Drive, 535 or 1822 Threading Machines
  • Cam-type feed actuation results in minimum operator effort to form roll grooves
  • Durable cast-iron housing withstands extensive field use
  • Depth adjustment screw separate from feed mechanism allows for consistent groove depth
  • Integral feed handle is part of overall unit
  • No extra tools required
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty