Would you like to log in again?
Weiler® Wide-Face Crimped Wire Wheel, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-03190 804-03090 804-03060 804-03200 804-03050 804-03240 804-03100 804-03000 804-03160 804-03590 804-03070 804-03010 804-03480 804-06645 804-03680 804-03150 804-03230 804-06655 804-03520 804-03030 804-03510 804-03140
300043-092EA 126.22 USD InStock
300043-092 300043-094 300044-114 300044-112 300043-096 300043-088 300043-098 300042-224 300042-772 300042-222 300043-090 300042-226 300041-814 300042-770 300042-220 300041-816 300041-624 300043-342 300043-100 300041-626 300043-102 300041-622
Weiler® Wide-Face Crimped Wire Wheel, ORS Nasco
Brushes
Consistent and effective performance for cleaning, light deburring, edge blending, roughening, and finishing.
  • Provides flexible brushing action
  • Maximum wire density and aggression, covers more surface area in a single pass
Designed for cleaning, light deburring, edge blending, roughening, and finishing. Use to remove rust, paint, oxides, and scale. Can be used with bench/pedestal grinders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty