Would you like to log in again?
Weiler® Hollow-End Knot Wire End Brush, ORS Nasco
Supplier: ORS Nasco
804-10217 804-10025 804-10141 804-10027 804-10030 804-10028 804-10208 804-10026 804-10029 804-10028P 804-10142 804-10031 804-10032
300087-842EA 54.44 USD InStock
300087-842 300046-684 300046-300 300046-686 300046-682 300046-298 300010-156 300010-157 300085-294 300010-158 300010-159 300047-446 300091-886
Weiler® Hollow-End Knot Wire End Brush, ORS Nasco
Brushes
Knot brushes are ideal for severe applications to remove rust, paint, oxides, and scale.
  • Hollow-end wire brush
  • Rugged knot-type construction provides aggressive, high-impact action for severe applications
  • Ideal for cleaning in recessed areas
Use with power drills, drill presses, die grinders, and right-angle air die grinders.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty