Would you like to log in again?
Type 29 Tiger Paw Angled Flap Discs, Weiler®
Supplier: Weiler
804-51121 804-51126 804-51125 804-51124 804-51119 804-51120
300053-352CS 91.22 USD InStock
300053-352 300053-350 300053-360 300053-358 300053-356 300053-354
Type 29 Tiger Paw Angled Flap Discs, Weiler®
Abrasives
  • 100% zirconia alumina grain is fast cutting with aggressive action and long life, resulting in fewer changeovers and increased productivity
  • Engineered poly/cotton cloth features excellent grain retention for maximum disc life
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty