Would you like to log in again?
Weiler® Crimped Wire Radial Wheel Brush, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-17966 804-17957 804-17969 804-17953 804-17962
300085-308PK 228.51 USD InStock
300085-308 300053-318 300085-310 300085-313 300085-306
Weiler® Crimped Wire Radial Wheel Brush, ORS Nasco
Brushes
Short trim length allows for more aggressive brushing action.
Designed for cleaning and finishing, rust and paint removal, and deburring. These brushes can be used with power drills, die grinders, and right-angle air die grinders. Use to remove rust, paint, oxide, and scale.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty