Would you like to log in again?
Weiler® Vortec Pro® Knot Wire Wheel, ORS Nasco
Supplier: ORS Nasco
Weiler®
804-36215 804-36298 804-36057 804-36212 804-36226 804-36257 804-36224 804-36022 804-36219 804-36213 804-36221 804-36293 804-36024 804-36028 804-36258 804-36294 804-36026 804-36058 804-36021 804-36013 804-36055 804-36297 804-36290 804-36291 804-36225 804-36279 804-36018 804-36217 804-36016 804-36278 804-36227 804-36027 804-36228 804-36218 804-36012 804-36255 804-36017 804-36295 804-36023 804-36216 804-36025 804-36015 804-36296
300085-518CS 194.66 USD InStock
300085-518 300085-616 300085-418 300085-618 300085-576 300085-630 300085-498 300085-432 300085-578 300085-434 300085-514 300085-414 300085-436 300085-516 300085-438 300085-416 300085-580 300085-440 300085-442 300085-420 300086-410 300085-628 300085-528 300085-620 300085-422 300085-520 300085-622 300085-424 300085-624 300085-524 300085-428 300085-626 300085-526 300085-592 300085-594 300085-494 300085-530 300085-496 300085-430 300091-942 300086-408 300091-960 300086-406
Weiler® Vortec Pro® Knot Wire Wheel, ORS Nasco
Brushes
Suitable for cleaning root and hot passes.
  • Maximum brushing action
  • For hard-to-reach areas, primarily for weld cleaning
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty