Would you like to log in again?
Vortec Pro Abrasive Flap Discs, Weiler®
Supplier: Weiler
804-31421 804-31344 804-31359 804-31402 804-31346 804-31419 804-31369 804-31363 804-31391 804-31316 804-31350 804-31422 804-31341 804-31309 804-31404 804-31408 804-31387 804-31349 804-31407 804-31401 804-31345 804-31416 804-31343 804-31389 804-31362 804-31351 804-31360 804-31414 804-31420 804-31352 804-31368 804-31370 804-31356 804-31388 804-31390 804-31392
300046-310EA 6.64 USD InStock
300046-310 300090-066 300044-466 300044-464 300090-082 300085-378 300085-356 300046-936 300085-358 300085-380 300085-382 300085-360 300085-384 300085-362 300085-364 300085-386 300045-926 300045-924 300010-209 300045-922 300090-074 300090-072 300046-308 300085-366 300085-368 300090-056 300010-210 300085-370 300010-211 300010-212 300085-374 300085-352 300085-354 300044-284 300044-282 300044-280
Vortec Pro Abrasive Flap Discs, Weiler®
Abrasives
Zirconium flap discs for use on steel, most metals, cast iron or non-ferrous soft metals.
  • High cut rate and smooth grinding action
  • Phenolic backing prevents disc from scratching the workpiece and offers durability and reinforcement
Ideal for grinding, finishing, weld blending or deburring.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty