Would you like to log in again?
KIMBLE® RAY-SORB® Amber Separatory Funnels, DWK Life Sciences
Supplier: DWK Life Sciences
KIMBLE® RAY-SORB®
29052F-125 29052F-500 29052F-250
30353-010EA 342.51 USD InStock
30353-010 30353-054 30353-032
KIMBLE® RAY-SORB® Amber Separatory Funnels, DWK Life Sciences
Funnels Separating Funnels
Designed with large lower stem I.D
for more complete separation. RAY-SORB® processed to protect materials sensitive to light of shorter wavelengths. Color-coded stopper. Designed from ASTM Specification E1096, Type IV requirements.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty