Would you like to log in again?
Uncontrolled Electromantles, with Metal Case, Electrothermal
Supplier: Electrothermal
CMU5000/EX1 CMU2000/EX1 CMU3000/EX1 36003-16 36003-12 36003-00 36103-04 36003-04
33743-448EA 649.14 USD InStock
33743-448 33743-454 10752-362 33743-456 33743-450 33743-462 33743-440 33743-452
Uncontrolled Electromantles, with Metal Case, Electrothermal
Heaters Heating Mantles
CMU heating mantles are used in wet chemistry to heat liquids in round bottom flasks
Ideal for use in fume hoods or remote working applications. The device must be used with a controller; recommended controllers are the MC5 and MC242. Electromantles accommodate round bottom flasks in capacities between 50mL and 5000mL. Element allows temperatures up to 450°C (842°F). Indicator lamps are provided for power and heater operation. Replaceable insulated heater cartridges are available. The rugged, easy to clean, powder-coated aluminium casing ensures long life and durability. Non-skid feet and support clamps are included (support rods not included). Flexible coiled heating elements absorb shock, minimizing the risk of flask breakage. The heating element is attached to a rigid thermal insulating cartridge to ensure maximum heat transfer and support. Standard safety features include a grounded earth stainless steel screen covering the heating element and double fuses.
The metal case is designed to remain "cool-to-the-touch" when in operation. Electrothermal heating mantles have a grounded stainless steel earth screen covering the element and are double fused for added safety.
Certifications: CE marked.
Ordering information: Electromantles are supplied with a powder-coated aluminum case.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty