Would you like to log in again?
VWR® Advanced Coveralls
Supplier: VWR
VWR®
414004-298 414004-304 414004-297 89234-868 414004-294 89234-866 89234-864 414004-295 414004-296 414004-299 414004-303 414004-291 414004-293 414004-300 414004-301 414004-292
414004-304CS 297.49 USD InStock
414004-304 414004-303 414004-301 414004-300 414004-299 414004-298 414004-297 89234-866 89234-868 89234-864 414004-296 414004-295 414004-294 414004-293 414004-292 414004-291
VWR® Advanced Coveralls
Disposable Garments
Coveralls are manufactured from a specially formulated breathable microporous fabric that is exceptionally clean and durable. These low-linting disposable garments provide a fluid barrier and dry-particulate protection, as well as resistance to common laboratory chemicals. Appropriate for industrial, pharmaceutical, food processing, construction, and certain environmental cleanup applications.
  • Significant fluid and particulate barrier
  • Durable, lightweight fabric
  • Excellent breathability and water vapor transmission rate
  • Soft tactile feel
All coveralls feature inset sleeves, front zipper, and elastic cuffs, ankles, and back
Coveralls are rigorously tested and manufactured in an ISO Certified facility under stringent process controls to ensure that each garment meets exacting quality standards and performs to specification. Lot controlled and validated through independent lab testing.
Available with an elastic hood and attached boot covers designed to reduce the risk of slipping in wet or dry conditions. Coveralls without a hood have a tapered collar. Serged seams. These advanced protection coveralls are made of PP.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty