Would you like to log in again?
Nalgene® Closures for Dropper Bottles, Thermo Scientific
Supplier: Thermo Scientific
Nalgene®
312760-0000 312760-0040 312760-0050 312760-0010 312760-0060 312760-0020
46300-574CS 387.36 USD InStock
46300-574 46300-572 46300-570 73521-300 46300-568 46300-576
Nalgene® Closures for Dropper Bottles, Thermo Scientific
Bottles Dropping and Dispensing Bottles
Nalgene closures for dropper bottles fit 312750 and 312751 bottles and must be ordered with dispensing tip to complete Dropper Bottle System.
The closures are available in natural, white, yellow, green, red, and blue. Neck finish for 46300-568 and 46300-570 is 15-415.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty