Would you like to log in again?
Pure™ XLG Pipette Tips, Molecular BioProducts
Supplier: Molecular BioProducts
Pure™
3702 3701
46600-020CS 1015.67 USD InStock
46600-020 46600-022
Pure™ XLG Pipette Tips, Molecular BioProducts
Pipette Tips
The Pure XLG tips are ideal for larger, more viscous samples.
  • Sterile tips are certified RNase-, DNase-, DNA-, and pyrogen-free
  • Non-sterile tips are autoclavable
Features include extended tip length and large orifice. Designed specifically for the pipetting of fragile cells, viscous samples, and genomic DNA. Tips are compatible with Pipetman*, Eppendorf*, Socorex*, and Rainin* pipettors.
Ordering information: Tips are packaged 96 per tray, 8 trays per pack.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty