Would you like to log in again?
Ward's® Thermochemistry In The Bag Inquiry Demonstration Activities
Supplier: Avantor
WARD470013-548
470013-548EA 42.99 USD InStock
470013-548
Ward's® Thermochemistry In The Bag Inquiry Demonstration Activities
Educational Classroom Kits and Activities
A Powerful Introduction to the Concept of Thermochemistry
  • Make a game to see who can guess the hottest or coldest bag
  • Discover variables and design experiments to test them
Feel the heat and study energy changes in chemical reactions through hands-on sensation of the actual temperature changes. This demonstration, safely contained within a plastic ziplock bag, will generate student questions and experiment ideas. Will they want to collect temperature data, or change the ratios of the starting materials, or both? You and your students control the demonstration parameters. The complete Thermochemistry In the Bag kit demonstrates both the endothermic and exothermic processes. Endothermic In the Bag studies a simple endothermic process of dissolving an inorganic salt that results in a solution that is \<10°C. Exothermic In the Bag, studies an exothermic process of dissolving an inorganic salt that will heat the solution up to >70°C. Kits contains enough materials to repeat the demonstrations about 20 times.
Materials Included:
Chemical for reactions
heavy duty plastic bags
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty