Would you like to log in again?
Diffraction Grating Glasses
Supplier: RAINBOW SYMPHONY INC SE
01101
470015-970PK 13 USD InStock
470015-970
Diffraction Grating Glasses
Educational Materials Physics Educational Materials Optics Learning Activities
These glasses make the study of light and color fun and easy.
  • Double-Axis Glasses
  • Lenses Separate Light from Other Sources
  • Economical
13,500 lines per inch double-axis holographic grating lenses separate light from any source into its spectral components for study and analysis.
Glasses are priced inexpensively enough to outfit your entire class.
Packaging: One package contains six glasses.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty