Would you like to log in again?
Streak and Color Collection
Supplier: Avantor
WARD470016-522
470016-522EA 49.3 USD InStock
470016-522
Streak and Color Collection
Specimens Mineral Specimens Mineral Collections
The powder a mineral produces (streak) is sometimes a different color than the mineral first appears. The 9 samples in this set were carefully chosen to help clearly demonstrate this physical property. A streak plate is also included.
  • Illustrates difference between streak and color
  • Includes a streak plate
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty