Would you like to log in again?
Magnetic Fingerprinting Supplies
Supplier: THE LYNN PEAVEY COMPANY
55575 1-0175 511 1-4021-1 55577 55574 55571
470020-206CS 35.1 USD InStock
470020-206 470202-488 470024-636 470024-634 470024-632 470024-630 470002-524
Magnetic Fingerprinting Supplies
Educational Materials Forensics Educational Materials Forensics Supplies
Apply powder and dust away excess to highlight fingerprints
  • Available in multiple colors
  • Available in two different sizes
Depress the plunger to pick up the magnetic fingerprint powder, dust the surface containing the prints, and then pull up on the plunger again to expel the excess powder into its container.
The larger base of the deluxe applicator allows you to pick up five times more powder than the standard applicator. Length of each: 5¹/₂".
Dual-Use Magnetic Fingerprint Powder: Highlights prints on both dark and light surfaces. Black Magnetic Fingerprint Powder: For highlighting prints on most standard surfaces. Red Magnetic Fingerprint Powder: Fluoresces under UV light. High lights prints on both dark and light surfaces. Deluxe applicator available to U.S. customers only; similar product will ship elsewhere.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty