Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Fixed Right Angle Clamp Holders

Supplier: Shiv Dial Sud & Sons
CH12364A RCLBH2
470111-722CS 1420.25 USD InStock
470111-722 470111-724
Fixed Right Angle Clamp Holders
Clamps General Purpose Clamps
Perpendicular, V-Shaped Jaws.

  • Thumbscrews 180° apart
  • Hold support rods up to 3/4"" OD at a 90° angle.

This all brass clamp holder allows you to connect support rods together up to 19 mm in diameter.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty