Would you like to log in again?
Somso® Male Torso with Head and Open Back
Supplier: MARCUS SOMMER
AS 23/1
470165-202EA 5955.09 USD InStock
470165-202
Somso® Male Torso with Head and Open Back
Models Human Biology Models
Natural size, in SOMSO-Plast®. Separates into 20 parts: brain with arteries (4), eye with muscles and optic nerve, halves of the lung (2), heart (2), liver, stomach (2), small and large intestine (3), opening appendix, omentum, half of right kidney, half of bladder, torso. On a green base. Height: 86 cm.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty