Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Density Cubes with Case

Supplier: EISCO SCIENTIFIC LLC
PH0109A
470230-038EA 75.9 USD InStock
470230-038
Density Cubes with Case
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
Set of 9 cubes, 25 mm side. Supplied in wooden box.

  • Density Block set consists of 9 blocks
  • 1" square blocks
  • Materials included brass, iron, copper, aluminum, hardwood, softwood, nylon, PVC and acrylic.
  • Supplied in 5.75" x 5.75" x 1.25" wooden box with hinges and clasp.
  • Perfect for student studying mass, volume and density

This set consists of nine cubes for density investigation, 25 mm side. Set of 9 comprising brass, iron, copper, aluminum, hardwood, softwood, nylon, PVC and acrylic. Supplied in wooden box.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty