Would you like to log in again?
Disposable Micropipette Tips
Supplier: Edvotek
VWR®
521016 D592 521000 637-2 521014 2249 1903
470149-166CS 67.75 USD InStock
470149-166 470233-654 470149-174 470148-816 470159-486 470019-832
Disposable Micropipette Tips
Pipette Tips
High quality micropipette tips to meet any lab's needs.
  • Certified DNase, RNase and pyrogen free
  • Material: PP
  • Autoclavable
  • Designed to fit most standard micropipettes
All quantities are supplied in bulk bags.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty