Would you like to log in again?
Ward's® Professional Insect Net
Supplier: Shiv Dial Sud & Sons
Ward's®
B11235C/3
470308-231CS 527.35 USD InStock
470308-231
Ward's® Professional Insect Net
Animal Supplies Animal Nets
A favorite of collectors for over 50 years.
  • Component assembly
  • Crafted for multiple uses
  • Strong and durable
  • Nets are 550 micron (µm) mesh
This professional insect net package is just what you need as a teacher. This package is assembled for basic aerial sweep collection. The heavy-duty 29.5" metal handle is longer than the student version. The metal handle has a larger diameter for strength, a high-gloss finish, and marine brass connection system. The 12" steel ring connects to the metal handle using pins and a retaining ring. The net easily slides onto the ring using a muslin sleeve and can be easily changed for a different style of net.
Accessories information: Replacement rings and nets can also be seen on our 'Build Your Own Insect Net' page.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty