Would you like to log in again?
Analysis of Rutherford Scattering, Leybold Products
Supplier: KLINGER EDUCATION PROD CORP SE
C1.1.3.4
470311-222EA 15326.19 USD InStock
470311-222
Analysis of Rutherford Scattering, Leybold Products
Educational Materials Chemistry Educational Materials Atomic Structure Learning Activities
Leybold Chemistry Experiments.
  • Determination of molar mass
  • States of matter
  • Analysis of Rutherford scattering
  • Chemical reactions and stoichiometry
The fact that an atom is 'mostly empty' was confirmed by Rutherford, Geiger and Marsden in a momentous experiment. They let a parallel bundle of α-particles fall on a very thin sheet of gold foil. In so doing they found that the vast majority of α-particles pass through the gold foil almost without any deflection at all and only very few are deflected to any large degree. They concluded from this that atoms comprise a nearly massless, extended shell and a massive core concentrated practically at a point.
Experiment C1.1.3.4, carries out an observation with an Am-241-preparation in a vacuum chamber. Depending on the scatter angle ϑ the scatter rate N(ϑ) of the α-particles is measured with a Geiger-Müller tube.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty