Would you like to log in again?
Dropper Assemblies
Supplier: GSC INTERNATIONAL SE
407-5-GR 407-4-DZ 405-4D-GR 407-4-GR
470346-708CS 62.2 USD InStock
470346-708 470338-740 470346-714 470338-706
Dropper Assemblies
Pipettes Pasteur/Transfer Pipettes
We sell many different sizes of dropper assemblies, varying by size, capacity, and the size of threaded neck they are designed to fit.
  • 24/400 neck size
  • Glass pipette with rubber bulb and plastic cap
Straight-talking and practical, our 24/400 neck size glass pipette with rubber bulb and plastic cap is the reliable dropper assembly you need in your lab. This no-frills design features a 20/400 cap that gets the job done without fuss.
The clear, flint glass construction is durable, while the simple rubber bulb and plastic cap provide a secure, leak-proof seal. Whether you're measuring out precious samples or refilling reagents, this dropper assembly puts functionality first. No flashy features, just a solid, trustworthy tool to keep your lab operations running smoothly. Get back to work with this honest, hardworking dropper - no extra bells and whistles required.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty